This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957271_s1
See other 4930441N18RIK GE Assays ›
Gene Symbol:
4930441N18Rik
Gene Name:
RIKEN cDNA 4930441N18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957271_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930441N18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019607.1 - 1 355 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714909.1 - 1 356 Chr. 4 - 83023799 - 83024245 [+] on Build GRCm38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cerberus 1 homolog (Xenopus laevis) Cer1 Cerl,
Cerl1,
Cerr1,
cer-1
-131.2kb Anti-sense Chr. 4 - 82879909 - 82892647 on NCBI Build GRCm38
Downstream tetratricopeptide repeat domain 39B Ttc39b 1810054D07Rik,
9130422G05Rik,
AI450603,
AV103077,
AW215585
196.1kb Anti-sense Chr. 4 - 83220301 - 83324270 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0