This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957260_s1
See other 4930434O05RIK GE Assays ›
Gene Symbol:
4930434O05Rik
Gene Name:
RIKEN cDNA 4930434O05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957260_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930434O05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK019595.1 - 1 248 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714871.1 - 1 245 Chr. 6 - 142989612 - 142990002 [+] on Build GRCm38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1 St8sia1 9330109E03Rik,
GD3S,
Sia-T,
Siat8,
Siat8a
-80.0 Anti-sense Chr. 6 - 142821541 - 142989530 on NCBI Build GRCm38
Downstream C2 calcium-dependent domain containing 5 C2cd5 5730419I09Rik,
C030008B15Rik,
CDP138
20.9kb Anti-sense Chr. 6 - 143010920 - 143100603 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_02_of_03_v2 A_68_P24907744 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907751 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907745 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907747 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907748 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907753 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907749 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907756 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24907746 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105080008 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0