Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1447798_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1447798_at |
- |
- |
Mouse Genome 430 2.0 Array |
1447798_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1600000526.mm.1 |
NR_045799.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000526.mm.1 |
NR_045799.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000526.mm.1 |
TC1600000056.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1447798_PM_at |
- |
- |
Mouse Expression Set 430 |
1447798_at |
- |
- |
Murine Genome U74v2 Set |
167348_at |
- |
- |
Murine Genome U74v2 Set |
167348_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1447798_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1447798_at |
- |
- |