This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956937_m1
See other 1700123O21RIK GE Assays ›
Gene Symbol:
1700123O21Rik
Gene Name:
RIKEN cDNA 1700123O21 gene
Gene Aliases:
-
Chromosome Location:
Chr.16: 5975587 - 5998496 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956937_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700123O21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045799.1 2 - 3 327 90
GenBank mRNA AK006584.1 - 2 - 3 328 90
AK018984.1 - 2 - 3 329 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707361.1 - 2 - 3 324 Chr. 16 - 5975590 - 5998461 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
36902
Gm36902 -42.2kb Anti-sense Chr. 16 - 5946938 - 5951145 on NCBI Build GRCm38
Downstream transmembrane protein 114 Tmem114 4930511J11Rik,
Cldn26
2.4mb Anti-sense Chr. 16 - 8409276 - 8425136 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000056.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000525.mm.1 NR_045799.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000525.mm.1 NR_045799.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000525.mm.1 TC1600000056.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0