This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956890_s1
See other 1700019A23RIK GE Assays ›
Gene Symbol:
1700019A23Rik
Gene Name:
RIKEN cDNA 1700019A23 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956890_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700019A23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK018838.1 - 1 47 65
AK018890.1 - 1 47 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706936.1 - 1 47 Chr. 7 - 30539666 - 30539868 [+] on Build GRCm38 65
GenBankmRNA BY706215.1 - 1 47 Chr. 7 - 30539666 - 30539868 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Rho GTPase activating protein 33 Arhgap33 AW324378,
NOMA-GAP,
Snx26,
TCGAP
-4.6kb Anti-sense Chr. 7 - 30522226 - 30535092 on NCBI Build GRCm38
Downstream heat shock protein,
alpha-crystallin-related,
B6
Hspb6 AA387366,
Gm479,
Hsp20
12.8kb Sense Chr. 7 - 30552634 - 30555439 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 778538 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0