This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956856_s1
See other 1500032F14RIK GE Assays ›
Gene Symbol:
1500032F14Rik
Gene Name:
RIKEN cDNA 1500032F14 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956856_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1500032F14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK018780.1 - 1 654 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV031986.1 - 1 200 Chr. 19 - 4256960 - 4257227 [-] on Build GRCm38 79
GenBankmRNA AW120923.1 - 1 Chr. 19 - 4256949 - 4257501 [+] on Build GRCm38 79
GenBankmRNA CR515982.1 - 1 Chr. 19 - 4256952 - 4257568 [+] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC102631992 LOC102631992 Sense Chr. 19 - 4209779 - 4257669 on NCBI Build GRCm38
Downstream slingshot homolog 3 (Drosophila) Ssh3 BC028922,
SSH-3,
mSSH-3L
4.0kb Sense Chr. 19 - 4261668 - 4270276 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2348126 AK018780.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P1091449 AK018780.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Murine Genome U74v2 Set 135981_at - -
Murine Genome U74v2 Set 135981_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 852070 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101580037 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0