This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956793_s1
See other 9130016M20RIK GE Assays ›
Gene Symbol:
9130016M20Rik
Gene Name:
RIKEN cDNA 9130016M20 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956793_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9130016M20Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK018631.1 - 1 146 104
AK050221.1 - 1 1291 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY720812.1 - 1 146 Chr. 19 - 31951227 - 31952194 [+] on Build GRCm38 104
GenBankmRNA AI173450.1 - 1 Chr. 19 - 31951041 - 31951464 [-] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein kinase,
cGMP-dependent,
type I
Prkg1 AW125416,
CGKI,
Gm19690,
Prkg1b,
Prkgr1b
-185.9kb Anti-sense Chr. 19 - 30564487 - 31765304 on NCBI Build GRCm38
Downstream N-acylsphingosine amidohydrolase 2 Asah2 AI585898 30.2kb Anti-sense Chr. 19 - 31982597 - 32108233 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2278775 AK050221.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541354 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760068 - -
Whole Mouse Genome G4122A A_52_P303754 AK050221.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P303754 AK018631.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P303754 AA087373.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111653 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453397_PM_at - -
Mouse Genome 430 2.0 Array 1453397_PM_at - -
Mouse Genome 430 2.0 Array 1453397_at - -
Mouse Expression Set 430 1453397_PM_at - -
Mouse Expression Set 430 1453397_at - -
Murine Genome U74v2 Set 99341_r_at - -
Murine Genome U74v2 Set 99341_PM_r_at - -
Affymetrix HT Mouse Genome 430B Array 1453397_at - -
Affymetrix HT Mouse Genome 430B Array 1453397_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 595199 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0