This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956646_s1
See other SNHG4 GE Assays ›
Gene Symbol:
Snhg4
Gene Name:
small nucleolar RNA host gene 4
Gene Aliases:
Gm17457
Chromosome Location:
Chr.18: 35553410 - 35558316 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956646_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snhg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK004333.1 - 1 49 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream small nucleolar RNA,
H/ACA box 74A
Snora74a EG436583,
RNU19,
U19
-79.0 Sense Chr. 18 - 35557030 - 35557227 on NCBI Build GRCm38
Downstream matrin 3 Matr3 1110061A14Rik,
2810017I02Rik,
AI841759,
AW555618,
D030046F20Rik,
mKIAA0723
4.3kb Sense Chr. 18 - 35562158 - 35592045 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0