This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956628_s1
See other 1110055C04RIK GE Assays ›
Gene Symbol:
1110055C04Rik
Gene Name:
RIKEN cDNA 1110055C04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956628_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110055C04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK004262.1 - 1 445 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY704505.1 - 1 441 Chr. 11 - 60796310 - 60797241 [+] on Build GRCm38 96
GenBankmRNA BU055257.1 - 1 Chr. 11 - 60796245 - 60796880 [-] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Smith-Magenis syndrome chromosome region,
candidate 8 homolog (human)
Smcr8 2310076G09Rik,
AI642055,
D030073L15Rik
-8.0kb Sense Chr. 11 - 60777525 - 60788287 on NCBI Build GRCm38
Downstream dehydrogenase/reductase (SDR family) member 7B Dhrs7b BC003479,
C79874,
C80074,
PexRAP
33.4kb Sense Chr. 11 - 60830631 - 60858423 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0