This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956574_s1
See other 1110035E04RIK GE Assays ›
Gene Symbol:
1110035E04Rik
Gene Name:
RIKEN cDNA 1110035E04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956574_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110035E04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK004108.1 - 1 355 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY704351.1 - 1 355 Chr. 8 - 107312677 - 107313671 [+] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear factor of activated T cells 5 Nfat5 AI225870,
B130038B15Rik,
CAG-8,
CAG80,
NFATL1,
OREBP,
TonEBP,
mKIAA0827,
nfatz
Sense Chr. 8 - 107293000 - 107379517 on NCBI Build GRCm38
Downstream NAD(P)H dehydrogenase,
quinone 1
Nqo1 AV001255,
Dia4,
Dtd,
Nmo-1,
Nmo1,
Nmor1,
Ox-1,
Ox1,
Qr1
74.2kb Anti-sense Chr. 8 - 107388225 - 107403205 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2375905 AK004108.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P1050743 AK004108.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432114_PM_at - -
Mouse Genome 430 2.0 Array 1432114_PM_at - -
Mouse Genome 430 2.0 Array 1432114_at - -
Mouse Expression Set 430 1432114_at - -
Mouse Expression Set 430 1432114_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432114_at - -
Affymetrix HT Mouse Genome 430B Array 1432114_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0