This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956391_s1
See other 1110004M10RIK GE Assays ›
Gene Symbol:
1110004M10Rik
Gene Name:
RIKEN cDNA 1110004M10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956391_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110004M10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK003437.1 - 1 953 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI988999.1 - 1 455 Chr. 2 - 92294263 - 92294828 [+] on Build GRCm38 71
GenBankmRNA CK428978.1 - 1 453 Chr. 2 - 92294291 - 92294818 [+] on Build GRCm38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1955 Mir1955 mir-1955,
mmu-mir-1955
-101.7kb Sense Chr. 2 - 92191977 - 92192074 on NCBI Build GRCm38
Downstream glycosyltransferase-like 1B Gyltl1b 5730485C17Rik,
AI891893,
Large2,
Largel,
mKIAA4105
70.2kb Anti-sense Chr. 2 - 92365046 - 92371940 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432584_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454286_PM_at - -
Mouse Genome 430 2.0 Array 1454286_PM_at - -
Mouse Genome 430 2.0 Array 1432584_at - -
Mouse Genome 430 2.0 Array 1454286_at - -
Mouse Genome 430 2.0 Array 1432584_PM_at - -
Mouse Expression Set 430 1432584_at - -
Mouse Expression Set 430 1454286_at - -
Mouse Expression Set 430 1454286_PM_at - -
Mouse Expression Set 430 1432584_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432584_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454286_at - -
Affymetrix HT Mouse Genome 430B Array 1432584_at - -
Affymetrix HT Mouse Genome 430B Array 1454286_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100730398 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0