This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956371_s1
See other 1110003F10RIK GE Assays ›
Gene Symbol:
1110003F10Rik
Gene Name:
RIKEN cDNA 1110003F10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956371_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110003F10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK003351.1 - 1 50 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY703608.1 - 1 48 Chr. 5 - 65405752 - 65406519 [+] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein L9 Rpl9 -14.3kb Anti-sense Chr. 5 - 65388364 - 65391431 on NCBI Build GRCm38
Downstream UDP-glucose dehydrogenase Ugdh Udpgdh 6.7kb Anti-sense Chr. 5 - 65413207 - 65435842 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_51_P520421 AK003351.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106760025 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0