This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956288_s1
See other 0610040A22RIK GE Assays ›
Gene Symbol:
0610040A22Rik
Gene Name:
RIKEN cDNA 0610040A22 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956288_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 0610040A22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK002857.1 - 1 1055 118

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 9 (sodium/hydrogen exchanger),
member 3
Slc9a3 9030624O13Rik,
AI930210,
NHE-3,
NHE3
Anti-sense Chr. 13 - 74086254 - 74169442 on NCBI Build GRCm38
Downstream exocyst complex component 3 Exoc3 2810050O03Rik,
E430013E20Rik,
Sec6,
Sec6l1
47.8kb Sense Chr. 13 - 74169796 - 74208729 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520112 - -
Whole Mouse Genome G4122A A_51_P280657 AK002857.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123253 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432770_PM_at - -
Mouse Genome 430 2.0 Array 1432771_PM_at - -
Mouse Genome 430 2.0 Array 1432771_at - -
Mouse Genome 430 2.0 Array 1432770_PM_at - -
Mouse Genome 430 2.0 Array 1432770_at - -
Mouse Expression Set 430 1432770_at - -
Mouse Expression Set 430 1432771_at - -
Mouse Expression Set 430 1432771_PM_at - -
Mouse Expression Set 430 1432770_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432770_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432770_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 339748 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105690138 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0