This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03955822_s1
See other 4930406D14RIK GE Assays ›
Gene Symbol:
4930406D14Rik
Gene Name:
RIKEN cDNA 4930406D14 gene
Gene Aliases:
AU022823
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03955822_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930406D14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015104.1 - 1 1251 90
AK086320.1 - 1 90
AU022823.1 - 1 90
BG072046.2 - 1 90
BG084797.2 - 1 387 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB702654.1 - 1 435 Chr. 11 - 86950572 - 86951081 [-] on Build GRCm38 90
GenBankmRNA BB700862.1 - 1 407 Chr. 11 - 86950563 - 86951050 [-] on Build GRCm38 90
GenBankmRNA BE996271.1 - 1 Chr. 11 - 86950546 - 86950879 [+] on Build GRCm38 90
GenBankmRNA BB707371.1 - 1 326 Chr. 11 - 86950572 - 86950971 [-] on Build GRCm38 90
GenBankmRNA BM940973.1 - 1 250 Chr. 11 - 86950544 - 86950879 [-] on Build GRCm38 90
GenBankmRNA CJ204337.1 - 1 315 Chr. 11 - 86950563 - 86950959 [-] on Build GRCm38 90
GenBankmRNA BB704577.1 - 1 405 Chr. 11 - 86950572 - 86951045 [-] on Build GRCm38 91
GenBankmRNA BB662855.1 - 1 Chr. 11 - 86950374 - 86950997 [+] on Build GRCm38 90
GenBankmRNA BB702288.1 - 1 445 Chr. 11 - 86950564 - 86951089 [-] on Build GRCm38 90
GenBankmRNA AA824076.1 - 1 157 Chr. 11 - 86950444 - 86950801 [-] on Build GRCm38 90
GenBankmRNA BB701171.1 - 1 408 Chr. 11 - 86950564 - 86951052 [-] on Build GRCm38 90
GenBankmRNA AI616182.1 - 1 163 Chr. 11 - 86950497 - 86950807 [-] on Build GRCm38 90
GenBankmRNA AU046108.1 - 1 Chr. 11 - 86950570 - 86951014 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream yippee-like 2 (Drosophila) Ypel2 6430570G24,
E130113K08Rik
Sense Chr. 11 - 86936425 - 86993849 on NCBI Build GRCm38
Downstream glycerophosphodiester phosphodiesterase domain containing 1 Gdpd1 2610020H15Rik 81.9kb Sense Chr. 11 - 87033794 - 87074953 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2381921 AK086320.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102680184 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0