This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03955621_s1
See other 4921519G19RIK GE Assays ›
Gene Symbol:
4921519G19Rik
Gene Name:
RIKEN cDNA 4921519G19 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03955621_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4921519G19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014924.1 - 1 750 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714442.1 - 1 745 Chr. 10 - 42855709 - 42856679 [-] on Build GRCm38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SEC63-like (S. cerevisiae) Sec63 5730478J10Rik,
AI649014,
AW319215
-22.4kb Anti-sense Chr. 10 - 42761496 - 42832514 on NCBI Build GRCm38
Downstream sex comb on midleg-like 4 (Drosophila) Scml4 9330161D17,
mFLJ00197
3.6kb Anti-sense Chr. 10 - 42860317 - 42960782 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331058 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432947_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432948_PM_at - -
Mouse Genome 430 2.0 Array 1432948_at - -
Mouse Genome 430 2.0 Array 1432947_at - -
Mouse Genome 430 2.0 Array 1432947_PM_at - -
Mouse Genome 430 2.0 Array 1432948_PM_at - -
Mouse Expression Set 430 1432947_PM_at - -
Mouse Expression Set 430 1432948_at - -
Mouse Expression Set 430 1432948_PM_at - -
Mouse Expression Set 430 1432947_at - -
Affymetrix HT Mouse Genome 430B Array 1432947_at - -
Affymetrix HT Mouse Genome 430B Array 1432948_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432948_at - -
Affymetrix HT Mouse Genome 430B Array 1432947_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 843033 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0