This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03955425_s1
See other 4833423F13RIK GE Assays ›
Gene Symbol:
4833423F13Rik
Gene Name:
RIKEN cDNA 4833423F13 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03955425_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4833423F13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014756.1 - 1 950 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV250457.1 - 1 Chr. 16 - 23294914 - 23295151 [+] on Build GRCm38 79
GenBankmRNA AW763815.1 - 1 Chr. 16 - 23294744 - 23295156 [-] on Build GRCm38 79
GenBankmRNA AI449104.1 - 1 Chr. 16 - 23294658 - 23295156 [-] on Build GRCm38 79
GenBankmRNA BY670060.1 - 1 267 Chr. 16 - 23294749 - 23295156 [+] on Build GRCm38 79
GenBankmRNA AI594650.1 - 1 Chr. 16 - 23294806 - 23295154 [-] on Build GRCm38 79
GenBankmRNA AA710841.1 - 1 289 Chr. 16 - 23294727 - 23295131 [+] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream beta galactoside alpha 2,6 sialyltransferase 1 St6gal1 AW742324,
Siat1,
St6Gal-I,
St6gal,
St6galI
Sense Chr. 16 - 23224728 - 23360350 on NCBI Build GRCm38
Downstream receptor transporter protein 1 Rtp1 Gm1766,
Gm604
133.4kb Sense Chr. 16 - 23428601 - 23433960 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0