This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03955155_s1
See other 4733401D01RIK GE Assays ›
Gene Symbol:
4733401D01Rik
Gene Name:
RIKEN cDNA 4733401D01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03955155_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4733401D01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014629.1 - 1 945 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream synaptopodin 2-like Synpo2l 1110054M18Rik,
Chap
-4.3kb Sense Chr. 14 - 20658945 - 20668349 on NCBI Build GRCm38
Downstream Sec24 related gene family,
member C (S. cerevisiae)
Sec24c 2610204K03Rik,
mKIAA0079
74.0 Anti-sense Chr. 14 - 20674285 - 20694852 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2200628 AK014629.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44587 - -
Whole Mouse Genome G4122A A_52_P1138688 AK014629.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453845_PM_at - -
Mouse Genome 430 2.0 Array 1453845_PM_at - -
Mouse Genome 430 2.0 Array 1453845_at - -
Mouse Expression Set 430 1453845_at - -
Mouse Expression Set 430 1453845_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453845_at - -
Affymetrix HT Mouse Genome 430B Array 1453845_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 632533 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0