This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954857_s1
See other 3300002P09RIK GE Assays ›
Gene Symbol:
3300002P09Rik
Gene Name:
RIKEN cDNA 3300002P09 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954857_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 3300002P09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI430424.1 - 1 73 77
AK014377.1 - 1 450 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA W89885.1 - 57 Chr. - - [-] on Build 77
GenBankmRNA DT930115.1 - 1 57 Chr. 10 - 80401038 - 80401465 [+] on Build GRCm38 77
GenBankmRNA BX519930.1 - 1 74 Chr. 10 - 80401022 - 80401418 [+] on Build GRCm38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mex3 homolog D (C. elegans) Mex3d BC059858,
Rkhd1
-13.0kb Anti-sense Chr. 10 - 80380355 - 80387651 on NCBI Build GRCm38
Downstream ubiquinol-cytochrome c reductase,
complex III subunit XI
Uqcr11 0710008D09Rik,
AL022707,
Uqcr
1.3kb Anti-sense Chr. 10 - 80402997 - 80406821 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 AF053974.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 AK019882.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 AK041505.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 AK045295.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 AK146773.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 BC065136.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777407 NM_009302.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32583 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453863_PM_at - -
Mouse Genome 430 2.0 Array 1453863_at - -
Mouse Genome 430 2.0 Array 1453863_PM_at - -
Mouse Expression Set 430 1453863_at - -
Mouse Expression Set 430 1453863_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453863_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453863_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 352731 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header