This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954767_s1
See other ILDR2 GE Assays ›
Gene Symbol:
Ildr2
Gene Name:
immunoglobulin-like domain containing receptor 2
Gene Aliases:
2810478N18Rik, 3110063L10Rik, AI852300, D1Ertd471e, Dbsm1, ENSMUSG00000040612, Ll
Chromosome Location:
Chr.1: 166254094 - 166316832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954767_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ildr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164528.1 NP_001158000.1 10 8075 74
GenBank mRNA AK014234.1 - 1 245 74
BC035277.1 - 1 3999 74
FJ024494.1 - 8022 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF463882.1 - 1 Chr. 1 - 166316477 - 166316823 [-] on Build GRCm38 74
GenBankmRNA CJ260814.1 - 1 291 Chr. 1 - 166316357 - 166316823 [+] on Build GRCm38 74
GenBankmRNA CJ260307.1 - 1 270 Chr. 1 - 166316378 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ259278.1 - 1 286 Chr. 1 - 166316362 - 166316822 [+] on Build GRCm38 74
GenBankmRNA BF466987.1 - 1 228 Chr. 1 - 166316420 - 166316745 [+] on Build GRCm38 74
GenBankmRNA AI852300.1 - 1 Chr. 1 - 166316426 - 166316839 [-] on Build GRCm38 74
GenBankmRNA CJ260281.1 - 1 270 Chr. 1 - 166316378 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ260574.1 - 1 261 Chr. 1 - 166316387 - 166316822 [+] on Build GRCm38 74
GenBankmRNA BF462852.1 - 1 273 Chr. 1 - 166316380 - 166316821 [+] on Build GRCm38 74
GenBankmRNA CJ259078.1 - 1 280 Chr. 1 - 166316368 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ258786.1 - 1 317 Chr. 1 - 166316330 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ259660.1 - 1 290 Chr. 1 - 166316358 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ258800.1 - 1 320 Chr. 1 - 166316328 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ260766.1 - 1 284 Chr. 1 - 166316364 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ260465.1 - 1 126 Chr. 1 - 166316523 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ259343.1 - 1 280 Chr. 1 - 166316368 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CO042153.1 - 1 Chr. 1 - 166316229 - 166316823 [-] on Build GRCm38 74
GenBankmRNA CJ261033.1 - 1 263 Chr. 1 - 166316385 - 166316822 [+] on Build GRCm38 74
GenBankmRNA BU056600.1 - 1 296 Chr. 1 - 166316352 - 166316823 [+] on Build GRCm38 74
GenBankmRNA CJ263812.1 - 1 295 Chr. 1 - 166316354 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ258212.1 - 1 272 Chr. 1 - 166316376 - 166316822 [+] on Build GRCm38 74
GenBankmRNA CJ261207.1 - 1 292 Chr. 1 - 166316356 - 166316824 [+] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream maelstrom homolog (Drosophila) Mael 4933405K18Rik,
AU019877
-77.7kb Anti-sense Chr. 1 - 166201385 - 166238744 on NCBI Build GRCm38
Downstream transcriptional adaptor 1 Tada1 2900026B15Rik,
D1Ertd251e,
Staf42,
Tada1l
62.3kb Sense Chr. 1 - 166379167 - 166393621 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

response to glucose
insulin secretion
cell differentiation
pancreas development
homeostasis of number of cells within a tissue

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024500.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024498.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024495.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 FJ024496.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952112 NM_001164528.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024500.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024498.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024495.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 FJ024496.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952112 NM_001164528.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00242195 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04342440 - -
Whole Mouse Genome G4122A A_51_P452533 NM_001164528.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P452533 AK014234.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P452533 FJ024494.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P221294 G49232.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138858 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00242195 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10351389 - -
Affymetrix Mouse Gene 1.0 ST Array 10351383 - -
Affymetrix Mouse Gene 1.0 ST Array 10351385 - -
Affymetrix Mouse Gene 1.0 ST Array 10359815 - -
Affymetrix Mouse Gene 1.0 ST Array 10351387 - -
Affymetrix Mouse Gene 1.0 ST Array 10351386 - -
Affymetrix Mouse Gene 1.0 ST Array 10351382 - -
Affymetrix Mouse Gene 1.0 ST Array 10351388 - -
Affymetrix Mouse Gene 1.0 ST Array 10351384 - -
Affymetrix HT MG-430 PM Array Plate 1436894_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436221_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436293_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10351380 - -
Affymetrix Mouse Gene 1.1 ST Array 10359814 - -
Mouse Genome 430 2.0 Array 1436894_PM_at - -
Mouse Genome 430 2.0 Array 1436894_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100039795_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100039795_at - -
Affymetrix Mouse Clariom S Array PSR0100012647.mm.1 NM_001164528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012647.mm.1 NM_001164528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012647.mm.1 TC0100001533.mm.1 MTA TC
Mouse Expression Set 430 1436894_at - -
Mouse Expression Set 430 1436894_PM_at - -
Murine Genome U74v2 Set 169845_PM_f_at - -
Murine Genome U74v2 Set 169845_f_at - -
Affymetrix HT Mouse Genome 430B Array 1436221_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1436221_at - -
Affymetrix HT Mouse Genome 430B Array 1436293_x_at - -
Affymetrix HT Mouse Genome 430B Array 1436293_PM_x_at - -
Affymetrix HT Mouse Genome 430B Array 1436894_at - -
Affymetrix HT Mouse Genome 430B Array 1436894_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 526904 - -
ABI Mouse Genome Survey Microarray 561161 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0