This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954624_s1
See other 3110037L02RIK GE Assays ›
Gene Symbol:
3110037L02Rik
Gene Name:
RIKEN cDNA 3110037L02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954624_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 3110037L02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014136.1 - 1 1346 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
29688
Gm29688 -215.5kb Anti-sense Chr. 8 - 126978041 - 126979494 on NCBI Build GRCm38
Downstream predicted gene,
32478
Gm32478 433.9kb Sense Chr. 8 - 127630400 - 127639705 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432619_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432618_PM_at - -
Mouse Genome 430 2.0 Array 1432618_PM_at - -
Mouse Genome 430 2.0 Array 1432618_at - -
Mouse Genome 430 2.0 Array 1432619_PM_at - -
Mouse Genome 430 2.0 Array 1432619_at - -
Mouse Expression Set 430 1432618_at - -
Mouse Expression Set 430 1432619_at - -
Mouse Expression Set 430 1432618_PM_at - -
Mouse Expression Set 430 1432619_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432618_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432619_at - -
Affymetrix HT Mouse Genome 430B Array 1432619_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432618_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100110594 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0