This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954416_s1
See other 3021401L19RIK GE Assays ›
Gene Symbol:
3021401L19Rik
Gene Name:
RIKEN cDNA 3021401L19 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954416_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 3021401L19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013907.1 - 1 651 113
AK034235.1 - 1 1698 113
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB081053.2 - 1 233 Chr. 2 - 139673241 - 139673919 [-] on Build GRCm38 113
GenBankmRNA BQ177272.1 - 1 611 Chr. 2 - 139673600 - 139674303 [-] on Build GRCm38 113

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream serine palmitoyltransferase,
long chain base subunit 3
Sptlc3 C130053K05Rik -35.7kb Anti-sense Chr. 2 - 139493913 - 139637674 on NCBI Build GRCm38
Downstream isthmin 1 homolog (zebrafish) Ism1 5430433G21Rik,
Ism
663.0 Anti-sense Chr. 2 - 139675007 - 139758581 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2189858 AK034235.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136535 - -
Whole Mouse Genome G4122A A_52_P647023 AK013907.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P647023 AK034235.1 mouse_transcriptome
Agilent Six_ChIP_01_of_03_v2 A_68_P21680261 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680272 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680273 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680266 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680260 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680265 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01838541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21680269 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 382922 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100130064 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0