This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954282_s1
See other 2900075N08RIK GE Assays ›
Gene Symbol:
2900075N08Rik
Gene Name:
RIKEN cDNA 2900075N08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954282_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2900075N08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013791.1 - 1 251 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY712755.1 - 1 249 Chr. X - 101322674 - 101323649 [+] on Build GRCm38 65
GenBankmRNA BY089753.1 - 1 55 Chr. X - 101322868 - 101323238 [+] on Build GRCm38 65
GenBankmRNA BU558768.1 - 1 109 Chr. X - 101322816 - 101323539 [+] on Build GRCm38 65
GenBankmRNA CX243135.1 - 1 534 Chr. X - 101322385 - 101323134 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neuroligin 3 Nlgn3 A230085M13Rik,
HNL3,
NL3
-1.3kb Sense Chr. X - 101299179 - 101321350 on NCBI Build GRCm38
Downstream gap junction protein,
beta 1
Gjb1 Cnx32,
Cx32
52.2kb Sense Chr. X - 101376376 - 101385629 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 760604 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header