This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953838_s1
See other 2900017F05RIK GE Assays ›
Gene Symbol:
2900017F05Rik
Gene Name:
RIKEN cDNA 2900017F05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953838_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2900017F05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013544.1 - 1 1044 113
BG072429.2 - 1 113
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU042656.1 - 1 Chr. 19 - 22777740 - 22778327 [-] on Build GRCm38 113

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transient receptor potential cation channel,
subfamily M,
member 3
Trpm3 6330504P12Rik,
9330180E14,
AU018608,
B930001P07Rik,
LTRPC3,
MLSN2
Sense Chr. 19 - 22138181 - 22989897 on NCBI Build GRCm38
Downstream RIKEN cDNA C330002G04 gene C330002G04Rik 218.6kb Anti-sense Chr. 19 - 22996890 - 23075906 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P706013 AK013544.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P706013 BG072429.2 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430096_PM_at - -
Mouse Genome 430 2.0 Array 1430096_PM_at - -
Mouse Genome 430 2.0 Array 1430096_at - -
Mouse Expression Set 430 1430096_at - -
Mouse Expression Set 430 1430096_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430096_at - -
Affymetrix HT Mouse Genome 430B Array 1430096_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 627103 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0