This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953325_s1
See other METTL1 GE Assays ›
Gene Symbol:
Mettl1
Gene Name:
methyltransferase like 1
Gene Aliases:
2810012D02Rik
Chromosome Location:
Chr.10: 127041643 - 127046365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953325_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK012721.1 - 1 353 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA547711.1 - 1 294 Chr. 10 - 127045931 - 127046439 [+] on Build GRCm38 95
GenBankmRNA AW457134.1 - 1 Chr. 10 - 127046010 - 127046359 [-] on Build GRCm38 94
GenBankmRNA AI838750.1 - 1 Chr. 10 - 127045892 - 127046365 [-] on Build GRCm38 95
GenBankmRNA BY711843.1 - 1 353 Chr. 10 - 127045871 - 127046363 [+] on Build GRCm38 95
GenBankmRNA CJ303604.1 - 1 263 Chr. 10 - 127045961 - 127046364 [+] on Build GRCm38 95
GenBankmRNA BU756283.1 - 1 Chr. 10 - 127046150 - 127046365 [-] on Build GRCm38 95
GenBankmRNA BU756791.1 - 1 Chr. 10 - 127046085 - 127046365 [-] on Build GRCm38 95
GenBankmRNA BM508316.1 - 1 Chr. 10 - 127046075 - 127046354 [-] on Build GRCm38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream methyltransferase like 21B Mettl21b -4.5kb Anti-sense Chr. 10 - 127032905 - 127041394 on NCBI Build GRCm38
Downstream cytochrome P450,
family 27,
subfamily b,
polypeptide 1
Cyp27b1 Cp2b,
Cyp1,
Cyp27b,
Cyp40,
P450c1,
Pddr,
Vdd1,
Vddr,
VddrI,
Vdr
1.9kb Sense Chr. 10 - 127048246 - 127053422 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0