This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953283_s1
See other C630043F03RIK GE Assays ›
Gene Symbol:
C630043F03Rik
Gene Name:
RIKEN cDNA C630043F03 gene
Gene Aliases:
AI851307
Chromosome Location:
Chr.4: 72201244 - 72203930 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953283_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C630043F03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027923.1 1 525 103
GenBank mRNA AK012513.1 - 1 449 103
AK039121.1 - 1 457 103
AK041409.1 - 1 527 103
AK042990.1 - 425 103
AK043880.1 - 1 517 103
AK053841.1 - 1 489 103
AK083285.1 - 1 446 103
AK083963.1 - 1 461 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB641636.1 - 1 516 Chr. 4 - 72201254 - 72201927 [+] on Build GRCm38 103
GenBankmRNA BB652975.1 - 1 446 Chr. 4 - 72201324 - 72202011 [+] on Build GRCm38 103
GenBankmRNA AW211099.1 - 1 478 Chr. 4 - 72201292 - 72201845 [+] on Build GRCm38 103
GenBankmRNA BB665543.1 - 1 486 Chr. 4 - 72201283 - 72201938 [+] on Build GRCm38 103
GenBankmRNA BB640250.1 - 1 423 Chr. 4 - 72201348 - 72202051 [+] on Build GRCm38 103
GenBankmRNA DV055542.1 - 449 Chr. - - [-] on Build 103
GenBankmRNA CB201524.1 - 1 413 Chr. 4 - 72201356 - 72202098 [+] on Build GRCm38 103
GenBankmRNA DV066411.1 - 1 515 Chr. 4 - 72201286 - 72202145 [+] on Build GRCm38 104
GenBankmRNA DV067447.1 - 1 482 Chr. 4 - 72201318 - 72202281 [+] on Build GRCm38 103
GenBankmRNA BQ565185.1 - 1 128 Chr. 4 - 72201642 - 72202129 [+] on Build GRCm38 103
GenBankmRNA BB632120.1 - 1 457 Chr. 4 - 72201313 - 72201957 [+] on Build GRCm38 103
GenBankmRNA AW989298.1 - 1 338 Chr. 4 - 72201431 - 72201936 [+] on Build GRCm38 103
GenBankmRNA BQ571368.1 - 1 361 Chr. 4 - 72201408 - 72202170 [+] on Build GRCm38 103
GenBankmRNA BQ551302.1 - 1 73 Chr. 4 - 72201696 - 72202307 [+] on Build GRCm38 103
GenBankmRNA BX512189.1 - 1 499 Chr. 4 - 72201291 - 72201853 [+] on Build GRCm38 103
GenBankmRNA BB656977.1 - 1 461 Chr. 4 - 72201309 - 72201935 [+] on Build GRCm38 103
GenBankmRNA BY682206.1 - 1 Chr. 4 - 72201708 - 72202095 [+] on Build GRCm38 103
GenBankmRNA BY711639.1 - 1 446 Chr. 4 - 72201324 - 72202096 [+] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transducin-like enhancer of split 1,
homolog of Drosophila E(spl)
Tle1 C230057C06Rik,
Estm14,
Grg-1,
Grg1,
Tle4l
-431.0 Anti-sense Chr. 4 - 72109945 - 72200893 on NCBI Build GRCm38
Downstream aldolase 1 A,
retrogene 1
Aldoart1 4921524E03Rik,
Aldo1-ps,
Aldo1-ps2,
Aldoa-ps2
648.5kb Anti-sense Chr. 4 - 72850583 - 72852634 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 AK004078.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 AK031866.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 AK080642.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 AK145197.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 BC038310.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 BC055847.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636286 NM_026812.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374384 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73203 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536973 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817276 BC019414.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817276 NM_134032.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601950 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88694 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658764 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31695 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161560 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161560 - -
Whole Mouse Genome G4122A A_51_P368743 AK053841.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P368743 AK043880.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P368743 AK039121.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P368743 AK041409.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P368743 NR_027923.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104107 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161560 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03309787 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23012987 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23012992 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03739209 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03877743 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23012995 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23012999 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433762_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1455542_PM_at - -
Mouse Genome 430 2.0 Array 1455542_at - -
Mouse Genome 430 2.0 Array 1433762_at - -
Mouse Genome 430 2.0 Array 1455542_PM_at - -
Mouse Genome 430 2.0 Array 1433762_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400005367.mm.1 NR_027923.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005367.mm.1 NR_027923.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005367.mm.1 TC0400000764.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400005364.mm.1 NR_027923.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005364.mm.1 NR_027923.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005364.mm.1 TC0400000764.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400005366.mm.1 NR_027923.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005366.mm.1 NR_027923.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005366.mm.1 TC0400000764.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400005369.mm.1 NR_027923.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005369.mm.1 NR_027923.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005369.mm.1 TC0400000764.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400005368.mm.1 NR_027923.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005368.mm.1 NR_027923.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400005368.mm.1 TC0400000764.mm.1 MTA TC
Mouse Expression Set 430 1455542_PM_at - -
Mouse Expression Set 430 1433762_PM_at - -
Mouse Expression Set 430 1433762_at - -
Mouse Expression Set 430 1455542_at - -
Affymetrix HT Mouse Genome 430B Array 1455542_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433762_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433762_at - -
Affymetrix HT Mouse Genome 430B Array 1455542_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 338254 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106510435 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0