This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953225_s1
See other 2700029L08RIK GE Assays ›
Gene Symbol:
2700029L08Rik
Gene Name:
RIKEN cDNA 2700029L08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953225_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2700029L08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK012303.1 - 1 554 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CAP-GLY domain containing linker protein 2 Clip2 B230327O20,
CLIP-115,
Clip1,
Cyln2,
WSCR4,
mKIAA0291,
wbscr4
Sense Chr. 5 - 134489383 - 134552468 on NCBI Build GRCm38
Downstream syncytin a Syna Gm453,
Gm52,
syncytin-A
36.5kb Sense Chr. 5 - 134557254 - 134577057 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430823_PM_at - -
Mouse Genome 430 2.0 Array 1430823_at - -
Mouse Genome 430 2.0 Array 1430823_PM_at - -
Mouse Expression Set 430 1430823_at - -
Mouse Expression Set 430 1430823_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430823_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430823_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0