This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953147_s1
See other 2610507I01RIK GE Assays ›
Gene Symbol:
2610507I01Rik
Gene Name:
RIKEN cDNA 2610507I01 gene
Gene Aliases:
5033414D05Rik
Chromosome Location:
Chr.11: 59197793 - 59202431 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953147_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2610507I01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037964.1 3 2355 71
GenBank mRNA AK012065.1 - 1 1251 71
AK162965.1 - 3 2310 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ312656.1 - 1 154 Chr. 11 - 59197917 - 59198497 [-] on Build GRCm38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream guanylate kinase 1 Guk1 AA409738,
AL033299,
AL033300,
AV026611,
Gmk,
Guk-1
-5.7kb Sense Chr. 11 - 59183855 - 59192207 on NCBI Build GRCm38
Downstream mitochondrial ribosomal protein L55 Mrpl55 2810038N09Rik,
L55mt,
MRP-L55
2.9kb Anti-sense Chr. 11 - 59202486 - 59206137 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2205650 AK162965.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2205650 NR_037964.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961553 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38511 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77272 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26926 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2205650 AK162965.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2205650 NR_037964.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02336910 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1431141_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1453749_PM_at - -
Mouse Genome 430 2.0 Array 1431141_at - -
Mouse Genome 430 2.0 Array 1453749_PM_at - -
Mouse Genome 430 2.0 Array 1453749_at - -
Mouse Genome 430 2.0 Array 1431141_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100025801.mm.1 NR_037964.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025801.mm.1 NR_037964.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025801.mm.1 TC1100002845.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100025802.mm.1 NR_037964.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025802.mm.1 NR_037964.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025802.mm.1 TC1100002845.mm.1 MTA TC
Mouse Expression Set 430 1431141_PM_at - -
Mouse Expression Set 430 1453749_at - -
Mouse Expression Set 430 1453749_PM_at - -
Mouse Expression Set 430 1431141_at - -
Affymetrix HT Mouse Genome 430B Array 1431141_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453749_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431141_at - -
Affymetrix HT Mouse Genome 430B Array 1453749_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 380086 - -
ABI Mouse Genome Survey Microarray 525897 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102470112 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0