This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953057_s1
See other 2610304O13RIK GE Assays ›
Gene Symbol:
2610304O13Rik
Gene Name:
RIKEN cDNA 2610304O13 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953057_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2610304O13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK011983.1 - 1 249 118
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY711177.1 - 1 249 Chr. 13 - 28725904 - 28726348 [-] on Build GRCm38 118

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 6368 Mir6368 Gm27800,
mmu-mir-6368
-15.0kb Sense Chr. 13 - 28710781 - 28710873 on NCBI Build GRCm38
Downstream predicted gene,
34639
Gm34639 163.2kb Sense Chr. 13 - 28889554 - 28898671 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_03_of_03_v2 A_68_P28907626 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907641 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11058948 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907631 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10617745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907634 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907637 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907620 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28907636 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100430215 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0