This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03952557_s1
See other 2310075M01RIK GE Assays ›
Gene Symbol:
2310075M01Rik
Gene Name:
RIKEN cDNA 2310075M01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03952557_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2310075M01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK010184.1 - 1 854 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear GTPase,
germinal center associated
Nuggc Gm600,
SLIP-GC
-98.9kb Sense Chr. 14 - 65593499 - 65648444 on NCBI Build GRCm38
Downstream PDZ binding kinase Pbk 2810434B10Rik,
AW538537,
D14Ertd732e,
TOPK
57.4kb Sense Chr. 14 - 65805911 - 65817822 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433248_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454524_PM_at - -
Mouse Genome 430 2.0 Array 1433248_PM_at - -
Mouse Genome 430 2.0 Array 1454524_PM_at - -
Mouse Genome 430 2.0 Array 1454524_at - -
Mouse Genome 430 2.0 Array 1433248_at - -
Mouse Expression Set 430 1454524_PM_at - -
Mouse Expression Set 430 1433248_at - -
Mouse Expression Set 430 1454524_at - -
Mouse Expression Set 430 1433248_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433248_at - -
Affymetrix HT Mouse Genome 430B Array 1454524_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454524_at - -
Affymetrix HT Mouse Genome 430B Array 1433248_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 625010 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0