This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03952433_s1
See other KLF9 GE Assays ›
Gene Symbol:
Klf9
Gene Name:
Kruppel-like factor 9
Gene Aliases:
2310051E17Rik, AA589643, BTEB-1, Bteb1, ENSMUSG00000055440, Gm9971
Chromosome Location:
Chr.19: 23141226 - 23166911 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03952433_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Klf9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK009928.1 - 652 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF119127.1 - 1 334 Chr. 19 - 23167428 - 23168122 [+] on Build GRCm38 65
GenBankmRNA CO039007.1 - 1 Chr. 19 - 23167370 - 23168129 [-] on Build GRCm38 65
GenBankmRNA AA204024.1 - 1 182 Chr. 19 - 23167594 - 23167910 [+] on Build GRCm38 65
GenBankmRNA AI303078.1 - 1 Chr. 19 - 23167662 - 23168129 [-] on Build GRCm38 65
GenBankmRNA AI848154.1 - 1 Chr. 19 - 23167645 - 23168129 [-] on Build GRCm38 65
GenBankmRNA AA959643.1 - 1 Chr. 19 - 23167644 - 23168127 [-] on Build GRCm38 65
GenBankmRNA BG147590.1 - 1 37 Chr. 19 - 23167725 - 23168054 [+] on Build GRCm38 65
GenBankmRNA BM933998.1 - 1 Chr. 19 - 23167568 - 23168134 [-] on Build GRCm38 65
GenBankmRNA BE331803.1 - 1 Chr. 19 - 23167690 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BY638460.1 - 1 46 Chr. 19 - 23167717 - 23168123 [+] on Build GRCm38 65
GenBankmRNA AW744676.1 - 1 Chr. 19 - 23167611 - 23168125 [-] on Build GRCm38 65
GenBankmRNA BE989496.1 - 1 Chr. 19 - 23167713 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BE134477.1 - 1 397 Chr. 19 - 23167365 - 23167956 [+] on Build GRCm38 65
GenBankmRNA BE989646.1 - 1 Chr. 19 - 23167708 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BQ177181.1 - 1 Chr. 19 - 23167454 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BF227892.1 - 1 Chr. 19 - 23167689 - 23168123 [-] on Build GRCm38 65
GenBankmRNA AA717754.1 - 1 201 Chr. 19 - 23167561 - 23168028 [+] on Build GRCm38 65
GenBankmRNA EH101976.1 - 1 Chr. 19 - 23167307 - 23167886 [-] on Build GRCm38 65
GenBankmRNA BE989967.1 - 1 Chr. 19 - 23167713 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BB767885.1 - 1 174 Chr. 19 - 23167587 - 23168123 [+] on Build GRCm38 65
GenBankmRNA BG229437.1 - 1 37 Chr. 19 - 23167725 - 23167987 [+] on Build GRCm38 65
GenBankmRNA CO428752.1 - 1 555 Chr. 19 - 23167207 - 23167861 [+] on Build GRCm38 65
GenBankmRNA BY709511.1 - 1 660 Chr. 19 - 23167111 - 23168080 [+] on Build GRCm38 65
GenBankmRNA AI197590.1 - 1 151 Chr. 19 - 23167611 - 23167914 [+] on Build GRCm38 65
GenBankmRNA BQ176403.1 - 1 Chr. 19 - 23167362 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BE990094.1 - 1 Chr. 19 - 23167638 - 23168129 [-] on Build GRCm38 65
GenBankmRNA DV044944.1 - 174 Chr. - - [-] on Build 65
GenBankmRNA AV484062.1 - 1 244 Chr. 19 - 23167525 - 23167897 [+] on Build GRCm38 65
GenBankmRNA BM565367.1 - 1 Chr. 19 - 23167529 - 23168129 [-] on Build GRCm38 65
GenBankmRNA AW912489.1 - 1 Chr. 19 - 23167662 - 23168127 [-] on Build GRCm38 65
GenBankmRNA AA444667.1 - 1 345 Chr. 19 - 23167418 - 23167847 [+] on Build GRCm38 65
GenBankmRNA CK337144.1 - 1 Chr. 19 - 23167545 - 23168126 [-] on Build GRCm38 65
GenBankmRNA AW990373.1 - 1 Chr. 19 - 23167652 - 23168126 [-] on Build GRCm38 65
GenBankmRNA BQ031740.1 - 1 Chr. 19 - 23167612 - 23168129 [-] on Build GRCm38 65
GenBankmRNA BE134043.1 - 1 Chr. 19 - 23167551 - 23168128 [-] on Build GRCm38 65
GenBankmRNA BU560537.1 - 226 Chr. - - [-] on Build 65
GenBankmRNA AI182243.1 - 1 83 Chr. 19 - 23167688 - 23168115 [+] on Build GRCm38 65
GenBankmRNA AI461709.1 - 1 Chr. 19 - 23167611 - 23168128 [-] on Build GRCm38 65
GenBankmRNA AA267992.1 - 1 199 Chr. 19 - 23167563 - 23168058 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Downstream structural maintenance of chromosomes 5 Smc5 MLZ-453,
SMC-5,
Smc5l1,
mKIAA0594,
mSMC5
38.5kb Anti-sense Chr. 19 - 23206441 - 23273908 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 339107 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0