This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03951251_s1
See other C230091D08RIK GE Assays ›
Gene Symbol:
C230091D08Rik Ube3a
Gene Name:
RIKEN cDNA C230091D08 gene
ubiquitin protein ligase E3A
Gene Aliases:
-
4732496B02, 5830462N02Rik, A130086L21Rik, Hpve6a, mKIAA4216
Chromosome Location:
Chr.7: 59307924 - 59324149 on Build GRCm38
Chromosome Location:
Chr.7: 59227814 - 59311536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm03951251_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C230091D08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK165737.1 - 1057 82
Gene Symbol: Ube3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC094605.1 - 1 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI466554.1 - 1 213 Chr. 7 - 59307708 - 59308167 [-] on Build GRCm38 82
GenBankmRNA CA888194.1 - 1 Chr. 7 - 59307823 - 59308432 [+] on Build GRCm38 82
GenBankmRNA CF742805.1 - 1 Chr. 7 - 59307465 - 59308193 [+] on Build GRCm38 82
GenBankmRNA CA891028.1 - 1 Chr. 7 - 59307699 - 59308203 [+] on Build GRCm38 82
GenBankmRNA CF910156.1 - 1 Chr. 7 - 59307881 - 59308529 [+] on Build GRCm38 82
GenBankmRNA CF908439.1 - 1 Chr. 7 - 59307886 - 59308462 [+] on Build GRCm38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATPase,
class V,
type 10A
Atp10a Atp10c,
pfatp
-477.7kb Anti-sense Chr. 7 - 58656143 - 58830035 on NCBI Build GRCm38
Downstream imprinted gene in the Prader-Willi syndrome region Ipw 311.0kb Sense Chr. 7 - 59619158 - 59678878 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0