This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03948821_s1
See other IL10RB GE Assays ›
Gene Symbol:
Il10rb
Gene Name:
interleukin 10 receptor, beta
Gene Aliases:
6620401D04Rik, AI528744, CRF2-4, Crfb4, D16H21S58, D21S58h, IL-10R2, Il10r2
Chromosome Location:
Chr.16: 91406235 - 91425834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03948821_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il10rb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK137922.1 - 1 1451 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB158216.1 - 1 213 Chr. 16 - 91409724 - 91410003 [+] on Build GRCm38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream interferon (alpha and beta) receptor 2 Ifnar2 AI747302,
Ifnar-2
-2.9kb Sense Chr. 16 - 91372215 - 91405589 on NCBI Build GRCm38
Downstream RIKEN cDNA A930006K02 gene A930006K02Rik 55.1kb Sense Chr. 16 - 91465104 - 91470123 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0