Assay Details
Gene Symbol: Nhsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136909.1 | - | 1 | 3752 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 28A | Ccdc28a |
1700009P13Rik, AI480677 |
-184.5kb | Anti-sense | Chr. 10 - 18213685 - 18234991 on NCBI Build GRCm38 |
Downstream | heme binding protein 2 | Hebp2 | SOUL | 116.8kb | Anti-sense | Chr. 10 - 18540123 - 18546076 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
