This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03948574_s1
See other B830008H07RIK GE Assays ›
Gene Symbol:
B830008H07Rik
Gene Name:
RIKEN cDNA B830008H07 gene
Gene Aliases:
AI662092
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03948574_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B830008H07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK046787.1 - 1 2229 96
AK135629.1 - 1 1549 96
BB333194.2 - 1 350 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA260921.1 - 1 46 Chr. 7 - 45471032 - 45471359 [-] on Build GRCm38 96
GenBankmRNA AV356187.2 - 1 304 Chr. 7 - 45470991 - 45471617 [-] on Build GRCm38 96
GenBankmRNA AI464801.1 - 1 46 Chr. 7 - 45470992 - 45471359 [-] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream BCL2-associated X protein Bax -4.1kb Sense Chr. 7 - 45461695 - 45466903 on NCBI Build GRCm38
Downstream dihydrodiol dehydrogenase (dimeric) Dhdh 1300018L09Rik,
AA591799,
Xld-1,
Xld1
701.0 Sense Chr. 7 - 45473563 - 45488873 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2209046 AK135629.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2209046 AI662092.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2209046 AK046787.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01507833 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119105 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1457791_PM_at - -
Mouse Genome 430 2.0 Array 1457791_at - -
Mouse Genome 430 2.0 Array 1457791_PM_at - -
Mouse Expression Set 430 1457791_PM_at - -
Mouse Expression Set 430 1457791_at - -
Murine Genome U74v2 Set 135691_PM_at - -
Murine Genome U74v2 Set 135691_at - -
Murine Genome U74v2 Set 137050_PM_at - -
Murine Genome U74v2 Set 137050_at - -
Affymetrix HT Mouse Genome 430B Array 1457791_at - -
Affymetrix HT Mouse Genome 430B Array 1457791_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0