This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03948106_s1
See other 1500015A07RIK GE Assays ›
Gene Symbol:
1500015A07Rik
Gene Name:
RIKEN cDNA 1500015A07 gene
Gene Aliases:
-
Chromosome Location:
Chr.18: 61726390 - 61728254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03948106_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1500015A07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029432.1 1 60 85
GenBank mRNA AK005240.1 - 1 61 85
AK133612.1 - 1 44 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB868961.1 - 1 44 Chr. 18 - 61726407 - 61726761 [+] on Build GRCm38 85
GenBankmRNA CJ162399.1 - 1 34 Chr. 18 - 61726416 - 61726870 [+] on Build GRCm38 85
GenBankmRNA BY705419.1 - 1 61 Chr. 18 - 61726390 - 61727303 [+] on Build GRCm38 85
GenBankmRNA BY241015.1 - 1 27 Chr. 18 - 61726423 - 61726898 [+] on Build GRCm38 85
GenBankmRNA BY777732.1 - 1 44 Chr. 18 - 61726407 - 61726777 [+] on Build GRCm38 85
GenBankmRNA BB617829.1 - 1 44 Chr. 18 - 61726407 - 61727018 [+] on Build GRCm38 85
GenBankmRNA BY105078.1 - 1 44 Chr. 18 - 61726407 - 61726806 [+] on Build GRCm38 85
GenBankmRNA BB845143.1 - 1 44 Chr. 18 - 61726407 - 61726830 [+] on Build GRCm38 85
GenBankmRNA BY085923.1 - 1 44 Chr. 18 - 61726407 - 61726784 [+] on Build GRCm38 85
GenBankmRNA BY772473.1 - 1 27 Chr. 18 - 61726423 - 61726803 [+] on Build GRCm38 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream GrpE-like 2,
mitochondrial
Grpel2 mt-GrpE#2,
mt-Grpel2
-46.0 Anti-sense Chr. 18 - 61712424 - 61726361 on NCBI Build GRCm38
Downstream actin filament associated protein 1-like 1 Afap1l1 AI173486 2.0kb Anti-sense Chr. 18 - 61730262 - 61786662 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2241204 BC047208.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2241204 AK133612.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2241204 NR_029432.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719082 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432783 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105016 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2241204 BC047208.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2241204 AK133612.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2241204 NR_029432.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105016 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100346 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105016 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1429162_PM_at - -
Mouse Genome 430 2.0 Array 1429162_at - -
Mouse Genome 430 2.0 Array 1429162_PM_at - -
Affymetrix Mouse Clariom S Array PSR1800004669.mm.1 NR_029432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004669.mm.1 NR_029432.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004669.mm.1 TC1800000637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004666.mm.1 NR_029432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004666.mm.1 NR_029432.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004666.mm.1 TC1800000637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004668.mm.1 NR_029432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004668.mm.1 NR_029432.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004668.mm.1 TC1800000637.mm.1 MTA TC
Mouse Expression Set 430 1429162_at - -
Mouse Expression Set 430 1429162_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429162_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429162_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 634283 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105720092 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header