This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03948004_m1
See other 4921515E04RIK GE Assays ›
Gene Symbol:
4921515E04Rik
Gene Name:
RIKEN cDNA 4921515E04 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 18078137 - 18369627 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03948004_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4921515E04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045711.1 11 - 12 2158 102
GenBank mRNA AK132982.1 - 11 - 12 2159 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV269390.2 - 4 - 5 525 Chr. 15 - 18078142 - 18165866 [-] on Build GRCm38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cadherin 9 Cdh9 -1.2mb Anti-sense Chr. 15 - 16728549 - 16857097 on NCBI Build GRCm38
Downstream RIKEN cDNA C030047K22 gene C030047K22Rik 617.0kb Sense Chr. 15 - 18697283 - 18818774 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001253.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2304786 AK132982.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707129 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756412 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546238 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380797 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205652 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747685 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55760 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436499 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356319 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895246 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175596 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693221 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776149 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290496 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787202 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455613 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462433 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427944 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277118 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546879 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786844 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163315 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638824 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788149 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205628 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934562 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700451 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720269 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291324 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521008 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476838 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834629 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293091 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170775 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767086 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821219 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794364 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840206 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529327 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558839 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646984 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467441 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228032 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941513 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392554 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462360 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44426 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747980 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906365 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188517 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872611 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727057 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969160 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586045 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179491 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127118 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225834 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722992 AF312019.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722992 AY028606.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722992 BC108971.2 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722992 NM_053249.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331934 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177949 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685426 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378573 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853761 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244131 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217928 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664856 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808778 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811546 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881927 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48847 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628792 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147561 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817223 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238952 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881581 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393940 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425993 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891348 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618670 AK006926.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618670 XM_001480489.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618670 XM_001480946.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295867 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616123 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112902 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116949 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140802 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665100 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828065 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341166 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588558 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797709 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51307 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551446 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838116 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26577 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28069 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714770 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577453 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415332 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914056 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474974 AK087787.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474974 NM_001081971.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199618 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536355 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132039 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30583 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50195 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79086 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171905 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102203 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52260 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163223 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144596 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102431 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119666 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72415 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121402 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160949 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14758 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79769 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66236 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298530 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298414 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298615 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298597 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298695 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298657 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298189 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02983927 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298276 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298519 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298567 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298364 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298494 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298545 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298295 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298218 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298466 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298438 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298643 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298257 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298332 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298149 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08298242 - -
Whole Mouse Genome G4122A A_52_P1018726 AK014898.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298189 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298567 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298332 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298414 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298530 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298242 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02983927 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298466 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08298657 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048414 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048403 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049153 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049167 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048397 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049172 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049134 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048406 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12336899 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049139 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048396 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049177 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049048 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049120 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048399 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048409 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049043 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049148 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049034 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048417 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12205416 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049159 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049030 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048413 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049138 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049037 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049013 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049158 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049020 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049141 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049123 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049039 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049011 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049140 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049165 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049035 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049049 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049131 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049157 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12253375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048405 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049129 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049122 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049124 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049151 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12217623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049033 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049031 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048408 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048415 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049066 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049027 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048419 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049018 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049175 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049133 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12355165 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049014 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12452538 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049126 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049178 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049006 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049169 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12121776 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049162 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049143 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12336889 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049170 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048391 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049017 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12253372 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049117 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049052 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049173 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12217624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049142 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049161 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049152 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049045 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048393 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049062 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048410 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049061 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049163 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30048402 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049016 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049067 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049053 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30049064 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453701_PM_at - -
Mouse Genome 430 2.0 Array 1453701_PM_at - -
Mouse Genome 430 2.0 Array 1453701_at - -
Affymetrix Mouse Clariom S Array PSR1500009942.mm.1 NR_045711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009942.mm.1 NR_045711.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009942.mm.1 TC1500001253.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500009943.mm.1 NR_045711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009943.mm.1 NR_045711.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009943.mm.1 TC1500001253.mm.1 MTA TC
Mouse Expression Set 430 1453701_at - -
Mouse Expression Set 430 1453701_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453701_at - -
Affymetrix HT Mouse Genome 430B Array 1453701_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 442920 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103850053 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0