This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947983_s1
See other BE691133 GE Assays ›
Gene Symbol:
BE691133
Gene Name:
expressed sequence BE691133
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947983_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BE691133
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK132899.1 - 1 952 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI558856.1 - 1 440 Chr. 4 - 137992016 - 137992774 [-] on Build GRCm38 65
GenBankmRNA AV265157.2 - 1 385 Chr. 4 - 137992121 - 137992721 [-] on Build GRCm38 65
GenBankmRNA BF472036.1 - 1 Chr. 4 - 137992005 - 137992623 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 4 gamma,
3
Eif4g3 1500002J22Rik,
4833436O05,
4930523M17Rik,
G1-419-52,
eIF4GII,
repro8
Anti-sense Chr. 4 - 137992525 - 138207081 on NCBI Build GRCm38
Downstream RIKEN cDNA 1700095J12 gene 1700095J12Rik 144.3kb Sense Chr. 4 - 138137580 - 138138518 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0