This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947962_m1
See other 4930404A12RIK GE Assays ›
Gene Symbol:
4930404A12Rik
Gene Name:
RIKEN cDNA 4930404A12 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947962_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930404A12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK132795.1 - 2 - 3 236 69
AK161424.1 - 2 - 3 233 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA464980.1 - 2 - 3 222 Chr. 5 - 124721998 - 124725064 [-] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATPase,
H+ transporting,
lysosomal V0 subunit A2
Atp6v0a2 8430408C20Rik,
AI385560,
ATP6a2,
AW489264,
Atp6n1d,
Atp6n2,
C76904,
ISF,
J6B7,
SHIF,
Stv1,
TJ6M,
TJ6s,
Tj6,
V-ATPase 116 kDa,
V-ATPase a2
Anti-sense Chr. 5 - 124712202 - 124724455 on NCBI Build GRCm38
Downstream dynein,
axonemal,
heavy chain 10
Dnah10 Dnahc10 1.3kb Anti-sense Chr. 5 - 124725018 - 124834827 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819591 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158934 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04964603 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24062548 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24062541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04123899 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04467225 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24062543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24062550 - -
Array Name Probe IDs Targets Target Type
Murine Genome U74v2 Set 169959_at - -
Murine Genome U74v2 Set 169959_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0