This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947961_m1
See other 4930404A12RIK GE Assays ›
Gene Symbol:
4930404A12Rik
Gene Name:
RIKEN cDNA 4930404A12 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947961_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930404A12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK132795.1 - 1 - 2 63 66
AK161424.1 - 1 - 2 60 66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR522711.1 - 2 - 3 Chr. 5 - 124721654 - 124725076 [+] on Build GRCm38 66
GenBankmRNA CA464980.1 - 1 - 2 49 Chr. 5 - 124721998 - 124725064 [-] on Build GRCm38 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream dynein,
axonemal,
heavy chain 10
Dnah10 Dnahc10 Anti-sense Chr. 5 - 124725018 - 124834827 on NCBI Build GRCm38
Downstream coiled-coil domain containing 92 Ccdc92 AW050178,
D5Bwg0834e
109.4kb Sense Chr. 5 - 124834432 - 124862221 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0