This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947646_m1
See other E530001K10RIK GE Assays ›
Gene Symbol:
E530001K10Rik
Gene Name:
RIKEN cDNA E530001K10 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 94160636 - 94264825 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947646_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E530001K10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK089118.1 - 1 - 2 240 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB559903.2 - 1 - 2 274 Chr. 2 - 94260966 - 94264758 [-] on Build GRCm38 99
GenBankmRNA BB609246.2 - 1 - 2 236 Chr. 2 - 94260964 - 94264755 [-] on Build GRCm38 99
GenBankmRNA BU557499.1 - 238 Chr. - - [-] on Build 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 670 Mir670 Mirn670,
mir-670,
mmu-mir-670
Sense Chr. 2 - 94261300 - 94261399 on NCBI Build GRCm38
Downstream tetratricopeptide repeat domain 17 Ttc17 9130020K17Rik,
D2Bwg1005e
36.0kb Sense Chr. 2 - 94300766 - 94406689 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106000 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121697 AK140370.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121701 AK140370.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00435449 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106000 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103773 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106000 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440120 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440126 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01811620 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02220582 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440138 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02220567 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01811631 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440118 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440129 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02220570 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440139 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02220572 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440114 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440133 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21440122 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02220581 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1444324_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1455275_PM_at - -
Mouse Genome 430 2.0 Array 1444324_at - -
Mouse Genome 430 2.0 Array 1455275_at - -
Mouse Genome 430 2.0 Array 1455275_PM_at - -
Mouse Genome 430 2.0 Array 1444324_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 414123_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 414123_at - -
Mouse Expression Set 430 1444324_at - -
Mouse Expression Set 430 1455275_PM_at - -
Mouse Expression Set 430 1444324_PM_at - -
Mouse Expression Set 430 1455275_at - -
Murine Genome U74v2 Set 166914_PM_i_at - -
Murine Genome U74v2 Set 166914_i_at - -
Affymetrix HT Mouse Genome 430B Array 1455275_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1455275_at - -
Affymetrix HT Mouse Genome 430B Array 1444324_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1444324_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0