This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947188_s1
See other CCL25 GE Assays ›
Gene Symbol:
Ccl25
Gene Name:
chemokine (C-C motif) ligand 25
Gene Aliases:
A130072A22Rik, AI852536, CKb15, Scya25, TECK
Chromosome Location:
Chr.8: 4325159 - 4360020 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947188_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccl25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK038019.1 - 1 646 80
AK086889.1 - 1 647 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB533386.2 - 1 112 Chr. 8 - 4325777 - 4326419 [+] on Build GRCm38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ELAV (embryonic lethal,
abnormal vision)-like 1 (Hu antigen R)
Elavl1 2410055N02Rik,
HUR,
Hua,
W91709
-133.0 Anti-sense Chr. 8 - 4284781 - 4325100 on NCBI Build GRCm38
Downstream ceramide synthase 4 Cers4 2900019C14Rik,
Lass4,
Trh1
166.6kb Sense Chr. 8 - 4492987 - 4526308 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0