This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03947170_s1
See other E030003N13RIK GE Assays ›
Gene Symbol:
E030003N13Rik
Gene Name:
RIKEN cDNA E030003N13 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03947170_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E030003N13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK086835.1 - 1 2253 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI451973.1 - 1 Chr. 10 - 117092662 - 117092915 [+] on Build GRCm38 89
GenBankmRNA BB529725.2 - 1 555 Chr. 10 - 117092663 - 117093345 [-] on Build GRCm38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chaperonin containing Tcp1,
subunit 2 (beta)
Cct2 Cctb -28.8kb Sense Chr. 10 - 117050997 - 117063814 on NCBI Build GRCm38
Downstream predicted gene,
40811
Gm40811 50.1kb Anti-sense Chr. 10 - 117145168 - 117149728 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P296006 AK086835.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P296006 AK051267.1 mouse_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0