This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03946573_s1
See other DHRS13OS GE Assays ›
Gene Symbol:
Dhrs13os
Gene Name:
dehydrogenase/reductase (SDR family) member 13, opposite strand
Gene Aliases:
A030003K02Rik, AA682085
Chromosome Location:
Chr.11: 78030304 - 78032498 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03946573_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs13os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK084880.1 - 1 757 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY214779.1 - 1 Chr. 11 - 78031773 - 78032136 [+] on Build GRCm38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PHD finger protein 12 Phf12 2410142K10Rik,
F630045O13,
PF1,
mKIAA1523
Anti-sense Chr. 11 - 77982754 - 78030539 on NCBI Build GRCm38
Downstream flotillin 2 Flot2 AI573412,
Esa,
reggie-2
5.3kb Anti-sense Chr. 11 - 78037868 - 78060434 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0