This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03946389_s1
See other D230034L24RIK GE Assays ›
Gene Symbol:
D230034L24Rik
Gene Name:
RIKEN cDNA D230034L24 gene
Gene Aliases:
AI664004
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03946389_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D230034L24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK084380.1 - 1 3750 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB470662.2 - 1 164 Chr. 16 - 72801390 - 72801999 [+] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream roundabout homolog 1 (Drosophila) Robo1 AW494633,
AW742721,
DUTT1,
Gm310
Sense Chr. 16 - 72027553 - 73047491 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930500H12 gene 4930500H12Rik 270.3kb Sense Chr. 16 - 73072346 - 73410430 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2418745 AK084380.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P715616 AK084380.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 388384 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header