This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03946107_s1
See other N4BP2L2 GE Assays ›
Gene Symbol:
N4bp2l2
Gene Name:
NEDD4 binding protein 2-like 2
Gene Aliases:
2700092H06Rik, zag1
Chromosome Location:
Chr.5: 150635973 - 150665612 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03946107_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N4bp2l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK083290.1 - 1 50 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI427170.1 - 1 Chr. 5 - 150642951 - 150650740 [+] on Build GRCm38 70
GenBankmRNA BB653674.1 - 1 51 Chr. 5 - 150642637 - 150643336 [-] on Build GRCm38 70
GenBankmRNA BY726563.1 - 1 70 Chr. 5 - 150642688 - 150643343 [-] on Build GRCm38 70
GenBankmRNA AI550581.1 - 3 364 Chr. 5 - 150643097 - 150661487 [-] on Build GRCm38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
38758
Gm38758 -30.5kb Sense Chr. 5 - 150599931 - 150610266 on NCBI Build GRCm38
Downstream PDS5,
regulator of cohesion maintenance,
homolog B (S. cerevisiae)
Pds5b AI646570,
AS3,
AW212954,
Aprin,
mKIAA0979
30.4kb Anti-sense Chr. 5 - 150673744 - 150810669 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0