Assay Details
Gene Symbol: 2610028E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015560.1 | 4 - 5 | 544 | 188 | ||
GenBank mRNA | AK011585.1 | - | 3 - 4 | 434 | 188 | |
AK031953.1 | - | 4 - 5 | 544 | 188 | ||
AK082619.1 | - | 4 - 5 | 469 | 188 | ||
AK083757.1 | - | 5 - 6 | 957 | 188 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
glutamate receptor, ionotropic, kainate 3 |
Grik3 |
9630027E11, GluK3, GluR7-3, Glur-7, Glur7 |
-177.3kb | Anti-sense | Chr. 4 - 125490709 - 125714173 on NCBI Build GRCm38 |
Downstream | colony stimulating factor 3 receptor (granulocyte) | Csf3r |
Cd114, Csfgr, G-CSF-R, G-CSFR |
130.2kb | Anti-sense | Chr. 4 - 126023441 - 126044975 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69857 | AK011585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69857 | AK011585.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553748 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677662 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781575 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163725 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477903 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751049 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59026 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121708 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94993 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10674 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69857 | AK011585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69857 | AK011585.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00980541 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05390704 | - | - |
Whole Mouse Genome G4122A | A_52_P69857 | AK011585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P69857 | AK011585.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110937 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00980541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03700825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439819 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03633770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03808499 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04043744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03343365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03484558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03778265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277102 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03553079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03939504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03364507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277099 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03760590 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03306219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04043738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03521345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03337782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04028739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03630818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03968046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03609140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04030187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03215688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03303347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04058536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03343368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03796860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03760588 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03760598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03337779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03941947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03215692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03457659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03941941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03279048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03941940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03687226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03703807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277062 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277143 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03306212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03306213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03633771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03279031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03233506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277127 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03326204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03484556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277218 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03364512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03633767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23276984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04031620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03619848 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23277010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1456708_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456708_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456708_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400028606.mm.1 | NR_015560.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028606.mm.1 | NR_015560.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028606.mm.1 | TC0400003525.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400028607.mm.1 | NR_015560.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028607.mm.1 | NR_015560.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028607.mm.1 | TC0400003525.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400028609.mm.1 | NR_015560.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028609.mm.1 | NR_015560.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028609.mm.1 | TC0400003525.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400028605.mm.1 | NR_015560.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028605.mm.1 | NR_015560.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400028605.mm.1 | TC0400003525.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456708_at | - | - |
Mouse Expression Set 430 | 1456708_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456708_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456708_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 468130 | - | - |
