This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03945806_s1
See other TMPPE GE Assays ›
Gene Symbol:
Tmppe
Gene Name:
transmembrane protein with metallophosphoesterase domain
Gene Aliases:
-
Chromosome Location:
Chr.9: 114401095 - 114411201 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03945806_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmppe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001200002.1 NP_001186931.1 2 6750 62
GenBank mRNA AK082033.1 - 1 1447 62
BE956926.1 - 1 62
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB709888.1 - 1 220 Chr. 9 - 114410876 - 114411202 [+] on Build GRCm38 62
GenBankmRNA CJ325705.1 - 1 358 Chr. 9 - 114410738 - 114411200 [+] on Build GRCm38 62
GenBankmRNA BB710616.1 - 1 331 Chr. 9 - 114410767 - 114411202 [+] on Build GRCm38 62
GenBankmRNA BM207740.2 - 1 Chr. 9 - 114410841 - 114411202 [-] on Build GRCm38 62
GenBankmRNA BB726152.1 - 1 334 Chr. 9 - 114410761 - 114411202 [+] on Build GRCm38 62
GenBankmRNA BQ559412.1 - 1 Chr. 9 - 114410911 - 114411202 [-] on Build GRCm38 62
GenBankmRNA CJ325661.1 - 1 382 Chr. 9 - 114410714 - 114411200 [+] on Build GRCm38 62
GenBankmRNA BY381771.1 - 1 347 Chr. 9 - 114410748 - 114411200 [+] on Build GRCm38 62
GenBankmRNA BE953874.1 - 1 386 Chr. 9 - 114410725 - 114411191 [+] on Build GRCm38 62
GenBankmRNA BB119588.1 - 1 203 Chr. 9 - 114410895 - 114411202 [+] on Build GRCm38 62
GenBankmRNA CB723442.1 - 1 140 Chr. 9 - 114410997 - 114411197 [+] on Build GRCm38 62
GenBankmRNA BB728872.1 - 1 325 Chr. 9 - 114410771 - 114411200 [+] on Build GRCm38 62
GenBankmRNA BB709880.1 - 1 215 Chr. 9 - 114410881 - 114411202 [+] on Build GRCm38 62
GenBankmRNA AW125515.1 - 1 Chr. 9 - 114410746 - 114411204 [-] on Build GRCm38 62
GenBankmRNA CJ325774.1 - 1 384 Chr. 9 - 114410713 - 114411200 [+] on Build GRCm38 62
GenBankmRNA DV656608.1 - 1 368 Chr. 9 - 114410728 - 114411175 [+] on Build GRCm38 62

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cartilage associated protein Crtap 5730529N23Rik,
CASP,
Leprel3,
P3h5
-18.9kb Anti-sense Chr. 9 - 114375131 - 114390712 on NCBI Build GRCm38
Downstream chemokine (C-C motif) receptor 4 Ccr4 C-C CKR-4,
CHEMR1,
Cmkbr4,
LESTR,
Sdf1r
79.1kb Anti-sense Chr. 9 - 114490316 - 114505473 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P679380 AK046107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679380 NM_001200002.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Mouse Genome 430 2.0 Array 1435795_at - -
Mouse Genome 430 2.0 Array 1435795_PM_at - -
Affymetrix Mouse Clariom S Array PSR0900012690.mm.1 NM_001200002.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012690.mm.1 NM_001200002.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012690.mm.1 TC0900001513.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012691.mm.1 NM_001200002.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012691.mm.1 NM_001200002.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012691.mm.1 TC0900001513.mm.1 MTA TC
Mouse Expression Set 430 1435795_at - -
Mouse Expression Set 430 1435795_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 354366 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0