This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03945801_s1
See other D930003E18RIK GE Assays ›
Gene Symbol:
D930003E18Rik
Gene Name:
RIKEN cDNA D930003E18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03945801_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D930003E18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK052957.1 - 1 1142 92
AK082011.1 - 1 848 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DT918079.1 - 1 148 Chr. 2 - 84838976 - 84839339 [-] on Build GRCm38 92
GenBankmRNA BB521769.2 - 1 359 Chr. 2 - 84838852 - 84839539 [-] on Build GRCm38 92
GenBankmRNA BQ268427.1 - 1 Chr. 2 - 84838850 - 84839446 [+] on Build GRCm38 92
GenBankmRNA W71563.1 - 218 Chr. - - [-] on Build 92
GenBankmRNA BB378464.2 - 1 409 Chr. 2 - 84838852 - 84839536 [-] on Build GRCm38 92
GenBankmRNA BQ268422.1 - 1 Chr. 2 - 84838850 - 84839407 [+] on Build GRCm38 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream translocase of inner mitochondrial membrane 10 Timm10 Tim13,
Timm13a
-8.6kb Anti-sense Chr. 2 - 84827021 - 84830213 on NCBI Build GRCm38
Downstream reticulon 4 receptor-like 2 Rtn4rl2 Ngr2,
Ngrh1,
Ngrl3
31.3kb Sense Chr. 2 - 84871309 - 84888507 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143228 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143228 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143228 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1440007_PM_at - -
Mouse Genome 430 2.0 Array 1440007_at - -
Mouse Genome 430 2.0 Array 1440007_PM_at - -
Mouse Expression Set 430 1440007_at - -
Mouse Expression Set 430 1440007_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1440007_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1440007_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0