This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03945754_s1
See other A130086K04RIK GE Assays ›
Gene Symbol:
A130086K04Rik
Gene Name:
RIKEN cDNA A130086K04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03945754_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A130086K04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK081662.1 - 1 252 107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB649222.1 - 1 Chr. 10 - 108921098 - 108921549 [-] on Build GRCm38 107

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream synaptotagmin I Syt1 AW124717,
G630098F17Rik,
SytI
Sense Chr. 10 - 108497650 - 109010975 on NCBI Build GRCm38
Downstream neuron navigator 3 Nav3 4732483H20Rik,
9630020C08Rik,
POMFIL1,
Pomfil1p,
mKIAA0938,
unc53H3
760.0kb Sense Chr. 10 - 109681573 - 110000391 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102810113 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0