This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03945269_s1
See other A530064N14RIK GE Assays ›
Gene Symbol:
A530064N14Rik
Gene Name:
RIKEN cDNA A530064N14 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03945269_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A530064N14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK080119.1 - 1 1257 73
BB221206.2 - 1 452 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI662606.1 - 1 Chr. 9 - 72688427 - 72688715 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 7265 Gm7265 EG639396 Sense Chr. 9 - 72686121 - 72696605 on NCBI Build GRCm38
Downstream protogenin homolog (Gallus gallus) Prtg A230098A12Rik,
Igdcc5
118.6kb Sense Chr. 9 - 72807274 - 72917307 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2328425 AK080119.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2328425 BB221206.2 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1445107_PM_at - -
Mouse Genome 430 2.0 Array 1445107_at - -
Mouse Genome 430 2.0 Array 1445107_PM_at - -
Mouse Expression Set 430 1445107_PM_at - -
Mouse Expression Set 430 1445107_at - -
Affymetrix HT Mouse Genome 430B Array 1445107_at - -
Affymetrix HT Mouse Genome 430B Array 1445107_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0